• USA Home
  • 61166 - Copper(I) bromide methyl sulfide complex

EMAIL THIS PAGE TO A FRIEND
61166 Sigma-Aldrich

Copper(I) bromide methyl sulfide complex

purum, ≥98.0% (RT)

Synonym: Cuprous bromide methyl sulfide complex

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Copper More...
InChI Key   PMHQVHHXPFUNSP-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (RT)
mp   132 °C (dec.)(lit.)

Description

Other Notes

Crystalline complex for the preparation of organocuprate reagents; review; their use for the preparation of trisubstituted olefins; Preparation of vinyl and allyl cuprates

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?