• USA Home
  • 61167 - Copper(II) carbonate basic

EMAIL THIS PAGE TO A FRIEND
61167 Sigma-Aldrich

Copper(II) carbonate basic

purum p.a., ≥95% (RT)

Synonym: Cupric carbonate basic

Purchase

Properties

Related Categories Copper Salts, Essential Chemicals, Research Essentials, Salts
grade   purum p.a.
assay   ≥95% (RT)
anion traces   chloride (Cl-): ≤1000 mg/kg
  sulfate (SO42-): ≤200 mg/kg
cation traces   Ca: ≤5000 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Fe: ≤100 mg/kg
  K: ≤100 mg/kg
  Na: ≤5000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤1000 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
GL6910000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?