• USA Home
  • 61257 - Cleaner concentrate for cuvette washing

EMAIL THIS PAGE TO A FRIEND
61257 Sigma-Aldrich

Cleaner concentrate for cuvette washing

alkaline

Synonym: Hellmanex III

Purchase

Description

Application

highly efficient, alkaline concentrate for the cleaning of cells made of glass and quartz

General description

Properties
Contains components for a trace-free decontamination without damaging the high precision polished surfaces. This product has no influence on enzymatic or serological reactions and has a low phosphate content.

Packaging

1 L in PE bottles
250 ML in glass bottles

Legal Information

Hellmanex is a trademark of Hellma GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?