EMAIL THIS PAGE TO A FRIEND
61321 Sigma-Aldrich

Lactobionic acid

purum

Synonym: 4-O-β-D-Galactopyranosyl-D-gluconic acid

  • CAS Number 96-82-2

  • Empirical Formula (Hill Notation) C12H22O12

  • Molecular Weight 358.30

  •  Beilstein Registry Number 95054

  •  EC Number 202-538-3

  •  MDL number MFCD00078147

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   JYTUSYBCFIZPBE-AMTLMPIISA-N
grade   purum
optical activity   [α]20/D +26±3°, 24 hr, c = 10% in H2O
ign. residue   ≤0.1%
loss   ≤5% loss on drying
mp   113-118 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?