• USA Home
  • 61520 - Lanthanum(III) nitrate hexahydrate

EMAIL THIS PAGE TO A FRIEND

61520 Sigma-Aldrich

Lanthanum(III) nitrate hexahydrate

puriss. p.a., ≥99.0% (titration)

Synonym: Nitric acid, lanthanum (III) salt, hexahydrate

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Catalysis and Inorganic Chemistry, Chemical Synthesis,
grade   puriss. p.a.
assay   ≥99.0% (titration)
form   crystalline
mp   65-68 °C
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  K: ≤10 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤10 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤10 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Lanthanum(III) nitrate hexahydrate along with aluminium acetylacetonate are dissolved in N,N-dimethylformamide to prepare lanthanum aluminate (LAO) thin films.† It used in the preparation of La0.8Sr0.2Co1-xFexO3 powder, perovskite type lanthanum calcium chromites titanates, LaF3 nanocrystals† and LnFeO3.† Preparation of bis(indolyl) methanes by the reaction of indoles with various aldehydes at room temperature in the presence of a catalytic amount of a La(NO3)3 · 6H2O afforded the corresponding bis(indolyl) methanes.† A wide variety of cyclic and acyclic dithioacetals have been prepared chemoselectively from their corresponding aldehydes with 1,3-dithiol and ethanethiol using catalytic amounts of lanthanum(III) nitrate hexahydrate as a catalyst at room temperature.†

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Explore the New Analytical Standards Portal

Ultra-high Purity Salts

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1477 5.1 / PGII
WGK Germany 
3
RTECS 
OE5250000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Synthesis of LaxAl2-xO3 Films using Ultrasonic Spray Pyrolysis Technique Meza RA, et al. ECS Transactions 33(3), 165-169, (2010)

Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. Kulangiappar KM, et al. Synth. Commun. 1(44), 2494-2502, (2014)

Lanthanum (III) nitrate hexahydrate catalyzed chemoselective thioacetalization of aldehydes Srinivasulu M, et al. J. Sulfur Chem. 28(3), 245-249, (2007)

Influence of Synthesis Parameters on Particle Size and Photoluminescence Characteristics of Ligand Capped Tb3+:LaF3 Gipson K, et al. Polymer 3, 2039-2052, (2011)

The Effect of the Lanthanide Ion-Type in LnFeO3 on the Catalytic Activity for the Hydrogen Evolution in Acidic Medium Atta NF, et al. Int. J. Electrochem. Sci. 9, 2132-2148, (2014)

CFTR and tight junctions in cultured bronchial epithelial cells. Nilsson HE, Dragomir A, Lazorova L, et al. Exp. Mol. Pathol. 88(1), 118-27, (2010)

Loading...


[Protective effects of La (NO3)3 on ryegrass seedlings photosynthetic apparatus under NaHCO3 stress]. Liu JX, Wang X, Wang RJ, et al. Ying Yong Sheng Tai Xue Bao 21(11), 2836-42, (2010)

Loading...


[Synthesis and characterization of La(III) binuclear complex with 2-hydroxy-l-naphthaldehyde-beta-alanine and its interaction with DNA]. Li YY, Fan YH, Bi CF, et al. Guang Pu Xue Yu Guang Pu Fen Xi 26(12), 2268-71, (2006)

Loading...


Epithelial and endothelial barriers in the olfactory region of the nasal cavity of the rat. Wolburg H, Wolburg-Buchholz K, Sam H, et al. Histochem. Cell Biol. 130(1), 127-40, (2008)

Loading...


Structural alterations of epididymal epithelial cells in cathepsin A-deficient mice affect the blood-epididymal barrier and lead to altered sperm motility. Hermo L, Korah N, Gregory M, et al. J. Androl. 28(5), 784-97, (2007)

Loading...


On-line arsenic co-precipitation on ethyl vinyl acetate turning-packed mini-column followed by hydride generation-ICP OES determination. Gil RA, Ferrúa N, Salonia JA, et al. J. Hazard. Mater. 143(1-2), 431-6, (2007)

Loading...


Penetration pathways induced by low-frequency sonophoresis with physical and chemical enhancers: iron oxide nanoparticles versus lanthanum nitrates. Lee SE, Choi KJ, Menon GK, et al. J. Invest. Dermatol. 130(4), 1063-72, (2010)

Loading...


Effect of some light rare earth elements on seed germination, seedling growth and antioxidant metabolism in Triticum durum. d'Aquino L, de Pinto MC, Nardi L, et al. Chemosphere 75(7), 900-5, (2009)

Loading...


Effects of diagnostic contrast-enhanced ultrasound on permeability of placental barrier: a primary study. Hua X, Zhu LP, Li R, et al. Placenta 30(9), 780-4, (2009)

Loading...


Synthesis of diamondoid lanthanide-polyoxometalate solids as tunable photoluminescent materials. Zhao WF, Zou C, Shi LX, et al. Dalton Trans. 41(33), 10091-6, (2012)

Loading...


Thalidomide treatment reduces the alteration of paracellular barrier function in mice ileum during experimental colitis. Mazzon E and Cuzzocrea S Shock 25(5), 515-21, (2006)

Loading...


Zona occludens-2 is critical for blood-testis barrier integrity and male fertility. Xu J, Anuar F, Ali SM, et al. Mol. Biol. Cell 20(20), 4268-77, (2009)

Loading...


Effect of electromagnetic pulse exposure on permeability of blood-testicle barrier in mice. Wang XW, Ding GR, Shi CH, et al. Biomed. Environ. Sci. 21(3), 218-21, (2008)

Loading...


Ambient particulate matter affects occludin distribution and increases alveolar transepithelial electrical conductance. Caraballo JC, Yshii C, Westphal W, et al. Respirology 16(2), 340-9, (2011)

Loading...


A PKC-beta inhibitor treatment reverses cardiac microvascular barrier dysfunction in diabetic rats. Wei L, Yin Z, Yuan Y, et al. Microvasc. Res. 80(1), 158-65, (2010)

Loading...


[Lanthanides induce neurotransmitter release from the vesicular pool in rat brain synaptosomes]. Lopatina LP, Vasim TV, Fedorovich SV, et al. Biofizika 50(6), 1120-4, (2005)

Loading...


Interactions between metal ions and carbohydrates. Syntheses and spectroscopic studies of several lanthanide nitrate-D-galactitol complexes. Yu L, Hua X, Pan Q, Yang L, et al. Carbohydr. Res. 346, 2278-2284, (2011)

Loading...


Permeability of the haemolymph-neural interface in the terrestrial snail Megalobulimus abbreviatus (Gastropoda, Pulmonata): an ultrastructural approach. Nóblega HG, Rigon F, Stenert C, et al. Comp. Biochem. Physiol. A. Mol. Integr. Physiol. 144(1), 119-24, (2006)

Loading...


Synthesis and spectroscopic properties of lanthanide nitrate complexes with a new amide-based quinoxaline-2,3-dione ligand. Song XQ, Wang YW, Zheng JR, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 68(3), 701-4, (2007)

Loading...


Effects of diagnostic ultrasound-targeted microbubble destruction on permeability of normal liver in rats. Yang D, Tan KB, Gao YH, et al. Ultrasonics 52(8), 1065-71, (2012)

Loading...


Optimisation of the chemical generation of singlet oxygen (1O2, 1Deltag) from the hydrogen peroxide-lanthanum(III) catalytic system using an improved NIR spectrometer. Pierlot C, Barbillat J, Nardello-Rataj V, et al. Photochem. Photobiol. Sci. 8(7), 1024-31, (2009)

Loading...


Effects of lanthanum on human lymphocytes viability and DNA strand break. Paiva AV, de Oliveira MS, Yunes SN, et al. Bull. Environ. Contam. Toxicol. 82(4), 423-7, (2009)

Loading...


Simulating transfer and persistence of a chemical marker powder for Lycopodium clavatum spores. Howarth J, Coulson S, and Newton A Forensic Sci. Int. 192(1-3), 72-7, (2009)

Loading...


Nanostructured La(Sr)CrO3 through gel combustion: sintering and electrical behavior. Nair SR, Purohit RD, Sinha PK, et al. J. Nanosci. Nanotechnol. 7(8), 2904-8, (2007)

Loading...


Synthesis, characterization and luminescent properties of lanthanide complexes with an unsymmetrical tripodal ligand. Yan ZZ, Dai GL, Liang HD, et al. Luminescence 26(3), 218-22, (2011)

Loading...


The role of chemical interactions between thorium, cerium, and lanthanum in lymphocyte toxicity. Oliveira MS, Duarte IM, Paiva AV, et al. Arch. Environ. Occup. Health 69(1), 40-5, (2014)

Loading...


Merck 14,5363

Fieser 11,283

Corp MSDS 1 (1), 2065:C / FT-IR 1 (2), 1242:C / RegBook 1 (3), 3415:F / Sax 6, 1684

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?