EMAIL THIS PAGE TO A FRIEND
61620 Sigma-Aldrich

Dodecanoic acid

purum, ≥96.0% (GC)

Synonym: Lauric acid, NSC 5026

  • CAS Number 143-07-7

  • Linear Formula CH3(CH2)10COOH

  • Molecular Weight 200.32

  •  Beilstein Registry Number 1099477

  •  EC Number 205-582-1

  •  MDL number MFCD00002736

  •  PubChem Substance ID 329759598

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids,
InChI Key   POULHZVOKOAJMA-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
mp   43-46 °C

Description

Biochem/physiol Actions

Substrate for CYP 4A11

Price and Availability


Laboratory Gloves

The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OE9800000
Flash Point(F) 
320 - 329 °F
Flash Point(C) 
160 - 165 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?