• USA Home
  • 61794 - Potassium chloride solution

EMAIL THIS PAGE TO A FRIEND
61794 Sigma-Aldrich

Potassium chloride solution

conductivity 11.67 mS/cm at 20 °C, analytical standard

Purchase

Properties

grade   analytical standard
InChI Key   WCUXLLCKKVVCTQ-UHFFFAOYSA-M
refractive index   n20/D 1.334
density   1.00 g/mL at 20 °C

Description

Application

Standard for the calibration of conductivity cells

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?