EMAIL THIS PAGE TO A FRIEND
61803 Sigma-Aldrich

2-Acetylpyridine

analytical standard

Synonym: Methyl 2-pyridyl ketone

  • CAS Number 1122-62-9

  • Empirical Formula (Hill Notation) C7H7NO

  • Molecular Weight 121.14

  •  Beilstein Registry Number 107759

  •  EC Number 214-355-6

  •  MDL number MFCD00006303

  •  PubChem Substance ID 329759687

Purchase

Properties

Related Categories A-B, A-C, A-F, A-L, Additional Standards,
grade   analytical standard
InChI Key   AJKVQEKCUACUMD-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.521(lit.)
  n20/D 1.522
bp   188-189 °C(lit.)
density   1.08 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
1
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?