EMAIL THIS PAGE TO A FRIEND
61825 Sigma-Aldrich

L-tert-Leucine

puriss., ≥99.0% (NT)

Synonym: (S)-2-Amino-3,3-dimethylbutyric acid, L-α-tert-Butylglycine

  • CAS Number 20859-02-3

  • Linear Formula (CH3)3CCH(NH2)CO2H

  • Molecular Weight 131.17

  •  Beilstein Registry Number 1721824

  •  MDL number MFCD00064218

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Leucine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   NPDBDJFLKKQMCM-SCSAIBSYSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D +7±1°, c = 5% in 5 M HCl
impurities   ~0.5% water
mp   ≥300 °C(lit.)

Description

Other Notes

This product has been replaced by 269107-ALDRICH | L-tert-Leucine 99%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

269107 L-tert-Leucine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?