• USA Home
  • 61855 - L-Leucinamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
61855 Sigma-Aldrich

L-Leucinamide hydrochloride

purum, ≥99.0% (AT)

  • CAS Number 10466-61-2

  • Linear Formula (CH3)2CHCH2CH(NH2)CONH2·HCl

  • Molecular Weight 166.65

  •  Beilstein Registry Number 4237021

  •  EC Number 233-952-2

  •  MDL number MFCD00013012

  •  PubChem Substance ID 329759698

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Leucine, Peptide Chemistry More...
InChI Key   VSPSRRBIXFUMOU-JEDNCBNOSA-N
grade   purum
assay   ≥99.0% (AT)
optical activity   [α]20/D +10±1°, c = 2% in H2O
optical purity   enantiomeric ratio: ≥99:1 (GC)
mp   245-248 °C
  254-256 °C(lit.)

Description

Other Notes

Chiral derivatizing agent for acids (as mixed anhydride) to determine the enantiomer ratio

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

286427 L-Leucinamide hydrochloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?