EMAIL THIS PAGE TO A FRIEND
62130 Sigma-Aldrich

(S)-(−)-Limonene

purum, ≥95.0% (sum of enantiomers, GC)

Synonym: (−)-p-Mentha-1,8-diene, (−)-Carvene, (S)-4-Isopropenyl-1-methyl cyclohexene

  • CAS Number 5989-54-8

  • Empirical Formula (Hill Notation) C10H16

  • Molecular Weight 136.23

  •  Beilstein Registry Number 2323991

  •  EC Number 227-815-6

  •  MDL number MFCD00001558

  •  PubChem Substance ID 329759711

Purchase

Properties

Related Categories Alkenes, Asymmetric Synthesis, Cell Biology, Chemical Synthesis, Chiral Building Blocks,
vapor density   4.7 (vs air)
vapor pressure   <3 mmHg ( 14.4 °C)
  1 mmHg ( 20 °C)
InChI Key   XMGQYMWWDOXHJM-SNVBAGLBSA-N
grade   purum
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D −90±5°, c = 10% in ethanol
expl. lim.   6.1 %
refractive index   n20/D 1.471(lit.)
  n20/D 1.473
bp   175-177 °C(lit.)
density   0.844 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by 218367-ALDRICH | (S)-(−)-Limonene 96%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2052 3 / PGIII
WGK Germany 
3
RTECS 
OS8350000
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?