• USA Home
  • 62136 - (+)-trans-Limonene 1,2-epoxide

EMAIL THIS PAGE TO A FRIEND
62136 Sigma-Aldrich

(+)-trans-Limonene 1,2-epoxide

analytical standard

Synonym: (+)-trans-p-Mentha-1,8-diene 1,2-epoxide, (1S,2R,4R)-4-Isopropenyl-1-methyl-1-cyclohexene 1,2-epoxide, (1S,4R)-1,2-Epoxy-p-menth-8-ene

  • CAS Number 6909-30-4

  • Empirical Formula (Hill Notation) C10H16O

  • Molecular Weight 152.23

  •  Beilstein Registry Number 80940

  •  MDL number MFCD16652609

  •  PubChem Substance ID 329759712

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Chromatography,
grade   analytical standard
InChI Key   CCEFMUBVSUDRLG-BBBLOLIVSA-N
assay   ≥95.0% (sum of enantiomers, GC)
form   neat
optical activity   [α]20/D +90±2°, neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.466
density   0.930 g/mL at 20 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
149 °F
Flash Point(C) 
65 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?