• USA Home
  • 62400 - Lithium acetylide ethylenediamine complex

EMAIL THIS PAGE TO A FRIEND
62400 Sigma-Aldrich

Lithium acetylide ethylenediamine complex

technical, ≥90% (T)

  • CAS Number 6867-30-7

  • Linear Formula LiC≡CH · NH2CH2CH2NH2

  • Molecular Weight 92.07

  •  Beilstein Registry Number 3593824

  •  EC Number 229-967-9

  •  MDL number MFCD00013183

  •  PubChem Substance ID 329759726

Purchase

Properties

Related Categories Alkynyl, Chemical Synthesis, Organolithium, Organometallic Reagents
InChI Key   QJQWXTYPTBEPGS-UHFFFAOYSA-N
grade   technical
assay   ≥90% (T)
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3131 8(4.3) / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

186155 Lithium acetylide, ethylenediamine complex, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?