• USA Home
  • 62465 - Lithium tert-butoxide

EMAIL THIS PAGE TO A FRIEND
62465 Sigma-Aldrich

Lithium tert-butoxide

technical, ≥95% (T)

  • CAS Number 1907-33-1

  • Linear Formula (CH3)3COLi

  • Molecular Weight 80.05

  •  Beilstein Registry Number 3620018

  •  EC Number 217-611-5

  •  MDL number MFCD00050479

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   LZWQNOHZMQIFBX-UHFFFAOYSA-N
grade   technical
assay   ≥95% (T)
impurities   ≤3% LiOH and Li2CO3

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3206 8(4.2) / PGII
WGK Germany 
3
RTECS 
UB8520000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

400173 Lithium tert-butoxide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?