• USA Home
  • 62483 - Lithium chloride solution

EMAIL THIS PAGE TO A FRIEND
62483 Sigma-Aldrich

Lithium chloride solution

1 M in glacial acetic acid

Purchase

Properties

Related Categories Analytical/Chromatography, Chemical Synthesis, Electrolytes, Inorganic Salts, Ion Sensor Materials,
InChI Key   KWGKDLIKAYFUFQ-UHFFFAOYSA-M
concentration   1 M in glacial acetic acid
refractive index   n20/D 1.381
density   1.08 g/mL at 20 °C

Description

Application

Filling solution for reference electrodes used for titrations in acetic acid.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2789PSN2 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?