EMAIL THIS PAGE TO A FRIEND
62551 Sigma-Aldrich

Lithium D-lactate

puriss., ≥99.0% (NT)

Synonym: (R)-2-Hydroxypropionic acid lithium salt, D-Lactic acid lithium salt

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acid Salts, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   GKQWYZBANWAFMQ-HSHFZTNMSA-M
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D +14.5±0.5°, c = 1% in H2O

Description

Other Notes

Stereoselective α-alkylation via dioxolanones

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

L1000 Lithium D-lactate, 95-100%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?