EMAIL THIS PAGE TO A FRIEND
62557 Sigma-Aldrich

Lithium ionophore I

Selectophore, function tested

Synonym: N,N′-Diheptyl-N,N′,5,5-tetramethyl-3,7-dioxanonanediamide, ETH 149

  • CAS Number 58821-96-8

  • Empirical Formula (Hill Notation) C25H50N2O4

  • Molecular Weight 442.68

  •  Beilstein Registry Number 2062796

  •  MDL number MFCD00042949

  •  PubChem Substance ID 329759736

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   WAPYYUBRXJPGKE-UHFFFAOYSA-N
quality   function tested

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Neutral ionophor for liquid-membrane electrodes of high selectivity for lithium ion;

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

62558 Lithium ionophore III, Selectophore, function tested
62559 Lithium ionophore II, Selectophore, function tested
99387 Lithium ionophore IV, Selectophore, function tested
62567 Lithium ionophore VI, Selectophore, function tested, ≥99.0% (HPLC)
62569 Lithium ionophore VII, Selectophore, function tested
62571 Lithium ionophore VIII, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?