• USA Home
  • 62569 - Lithium ionophore VII

EMAIL THIS PAGE TO A FRIEND
62569 Sigma-Aldrich

Lithium ionophore VII

Selectophore, function tested

Synonym: 6-Dodecyl(14-crown-4)-6-ethanol diethyl phosphate, 6-[2-(Diethylphosphonooxy)ethyl]-6-dodecyl-14-crown-4

  • CAS Number 106868-29-5

  • Empirical Formula (Hill Notation) C28H57O8P

  • Molecular Weight 552.72

  •  Beilstein Registry Number 6013865

  •  MDL number MFCD00037656

  •  PubChem Substance ID 24882246

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   WTJIVAYXSTYCNI-UHFFFAOYSA-N
quality   function tested

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Lithium ionophore possessing a high selectivity for Li+ against Na+ and K+ cations

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

solution in 0.1 mL heptane

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1206 - class 3 - PG 2 - Heptanes, solution
WGK Germany 
3
Flash Point(F) 
24.8 °F
Flash Point(C) 
-4 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

62557 Lithium ionophore I, Selectophore, function tested
62558 Lithium ionophore III, Selectophore, function tested
62559 Lithium ionophore II, Selectophore, function tested
99387 Lithium ionophore IV, Selectophore, function tested
62567 Lithium ionophore VI, Selectophore, function tested, ≥99.0% (HPLC)
62571 Lithium ionophore VIII, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?