EMAIL THIS PAGE TO A FRIEND
62607 Sigma-Aldrich

Lithium salicylate

purum, ≥98.0% (NT)

Synonym: Salicylic acid lithium salt

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
InChI Key   PSBOOKLOXQFNPZ-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (NT)
impurities   ≤0.5% water
mp   >350 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

518026 Lithium salicylate, 99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?