EMAIL THIS PAGE TO A FRIEND
62855 Sigma-Aldrich

L-Lysine monohydrate

purum, ≥98.0% (NT)

  • CAS Number 39665-12-8

  • Empirical Formula (Hill Notation) C6H14N2O2 · H2O

  • Molecular Weight 164.20

  •  Beilstein Registry Number 4349963

  •  MDL number MFCD00151035

  •  PubChem Substance ID 24882274

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Lysine, Peptide Chemistry More...
InChI Key   HZRUTVAFDWTKGD-JEDNCBNOSA-N
grade   purum
assay   ≥98.0% (NT)
optical activity   [α]20/D +22.5±1°, c = 2% in 6 M HCl

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

282677 L-Lysine hydrate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?