EMAIL THIS PAGE TO A FRIEND
62860 Sigma-Aldrich

DL-Lysine

purum, ≥95.0% (NT)

Synonym: (±)-2,6-Diaminocaproic acid

  • CAS Number 70-54-2

  • Linear Formula NH2(CH2)4CH(NH2)COOH

  • Molecular Weight 146.19

  •  Beilstein Registry Number 1616991

  •  EC Number 200-740-6

  •  MDL number MFCD00064432

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Lysine, Peptide Chemistry More...
InChI Key   KDXKERNSBIXSRK-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (NT)
impurities   ≤2% water
mp   170 °C (dec.)(lit.)
  ~175 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?