• USA Home
  • 63141 - Magnesium sulfate monohydrate

EMAIL THIS PAGE TO A FRIEND
63141 Sigma-Aldrich

Magnesium sulfate monohydrate

purum, ≥97.0% (KT)

Synonym: Kieserite

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Magnesium, Magnesium Salts,
InChI Key   LFCFXZHKDRJMNS-UHFFFAOYSA-L
grade   purum
assay   ≥97.0% (KT)
mp   150 °C (dec.)(lit.)
density   2.57 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OM4500000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

434183 Magnesium sulfate monohydrate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?