EMAIL THIS PAGE TO A FRIEND
63285 Sigma-Aldrich

N-Maleoyl-β-alanine

≥98.0% (HPLC)

Synonym: 3-Maleimidopropionic acid, N-(2-Carboxyethyl)maleimide

  • CAS Number 7423-55-4

  • Empirical Formula (Hill Notation) C7H7NO4

  • Molecular Weight 169.13

  •  Beilstein Registry Number 1528952

  •  MDL number MFCD00043030

  •  PubChem Substance ID 329759823

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Cyclic Imides, Organic Building Blocks More...
InChI Key   IUTPJBLLJJNPAJ-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
mp   103-106 °C(lit.)
  108-111 °C

Description

Other Notes

Reagent for the modification of peptides, proteins with an SH-label, probe for membrane SH-groups, Used for the preparation of peptide-protein conjugates

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

394815 N-Maleoyl-β-alanine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?