EMAIL THIS PAGE TO A FRIEND
63360 Sigma-Aldrich

Malonyl chloride

purum, ≥97.0% (AT)

Synonym: Malonyl dichloride

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   SXYFKXOFMCIXQW-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
refractive index   n20/D 1.465(lit.)
  n20/D 1.466
bp   53-55 °C/19 mmHg(lit.)
density   1.449 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
1
Flash Point(F) 
116.6 °F
Flash Point(C) 
47 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M1601 Malonyl chloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?