• USA Home
  • 63450 - D-(−)-Mandelic acid

EMAIL THIS PAGE TO A FRIEND
63450 Sigma-Aldrich

D-(−)-Mandelic acid

puriss., ≥99.0% (T)

Synonym: (R)-α-Hydroxyphenylacetic acid

  • CAS Number 611-71-2

  • Empirical Formula (Hill Notation) C8H8O3

  • Molecular Weight 152.15

  •  Beilstein Registry Number 2691094

  •  EC Number 210-276-6

  •  MDL number MFCD00064251

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   IWYDHOAUDWTVEP-SSDOTTSWSA-N
grade   puriss.
assay   ≥99.0% (T)
optical activity   [α]20/D −155±5°, c = 5% in H2O
optical purity   enantiomeric ratio: ≥99:1 (GC)
mp   130-133 °C

Description

Other Notes

Reagent used for the resolution of bases; Resolution of alcohols via esters; Stereoselective α-alkylation via dioxolanones

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
>374 °F
Flash Point(C) 
>190 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

154210 (R)-(−)-Mandelic acid, ReagentPlus®, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?