• USA Home
  • 63533 - Manganese standard for IC

EMAIL THIS PAGE TO A FRIEND
63533 Sigma-Aldrich

Manganese standard for IC

certified reference material, ready-to-use, traceable to BAM

Synonym: Manganese(II) nitrate solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Cationic Solutions, Chromatography,
grade   certified reference material
InChI Key   PWHULOQIROXLJO-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
contains   0.5 mM vitamin C as stabilizer
concentration   1.000 g/L in H2O
application(s)   ion chromatography: suitable
format   single component solution

Description

General description

filtered through a 0.45 μm membrane filter

Preparation Note

prepared with Mn(NO3)2 and H2O

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

51439 Manganese Standard for IC, TraceCERT®, 1000 mg/L Mn+2 in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?