• USA Home
  • 63541 - Manganese(0) carbonyl

EMAIL THIS PAGE TO A FRIEND
63541 Sigma-Aldrich

Manganese(0) carbonyl

purum, ≥98.0% Mn basis

Synonym: Decacarbonyl dimanganese(0), Dimanganese(0) decacarbonyl

Purchase

Properties

InChI Key   QFEOTYVTTQCYAZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% Mn basis
mp   154 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3288 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

245267 Manganese(0) carbonyl, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?