• USA Home
  • 63555 - Manganese(II) sulfate monohydrate

EMAIL THIS PAGE TO A FRIEND
63555 Sigma-Aldrich

Manganese(II) sulfate monohydrate

purum p.a., ≥98.0% (T)

Purchase

Properties

Related Categories Essential Chemicals, Manganese Salts, Research Essentials, Salts
InChI Key   ISPYRSDWRDQNSW-UHFFFAOYSA-L
grade   purum p.a.
assay   ≥98.0% (T)
anion traces   chloride (Cl-): ≤100 mg/kg
cation traces   Ca: ≤1000 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤5000 mg/kg
  Na: ≤2000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1
RTECS 
OP0893500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?