• USA Home
  • 63598 - Multi-element Ion Chromatography Standard solution II

EMAIL THIS PAGE TO A FRIEND
63598 Sigma-Aldrich

Multi-element Ion Chromatography Standard solution II

analytical standard, in H2O

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Anion Standards, Chromatography,
grade   analytical standard
shelf life   limited shelf life, expiry date on the label
composition   nitrate, 1.00 g/L
  phosphate, 1.00 g/L
  sulfate, 1.00 g/L
concentration   in H2O
application(s)   ion chromatography: suitable
format   multi-component solution

Description

General description

filtered through a 0.45 μm membrane filter

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Preparation Note

prepared with high purity salts and water

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?