EMAIL THIS PAGE TO A FRIEND
63658 Sigma-Aldrich

(+)-Menthol

puriss. p.a., terpene standard for GC, ≥99.0% (sum of enantiomers, GC)

Synonym: (1S,2R,5S)-2-Isopropyl-5-methylcyclohexanol

  • CAS Number 15356-60-2

  • Empirical Formula (Hill Notation) C10H20O

  • Molecular Weight 156.27

  •  Beilstein Registry Number 1902292

  •  EC Number 239-387-8

  •  MDL number MFCD00062983

  •  PubChem Substance ID 329759884

Purchase

Properties

Related Categories Additional Standards, Alcohols, Aliphatic, Analytical Standards, Analytical/Chromatography,
grade   terpene standard for GC
InChI Key   NOOLISFMXDJSKH-AEJSXWLSSA-N
grade   puriss. p.a.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +49±1°, c = 10% in ethanol
optical purity   enantiomeric ratio: ≥98.5:1.5 (GC)
shelf life   limited shelf life, expiry date on the label
mp   ~43 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
OT0700000
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Menthol Enantiomers on α-DEX™ 120

From our library of Articles, Sigma-Aldrich presents GC Analysis of Menthol Enantiomers on α-DEX™ 120
Keywords: Chromatography, Cosmetics, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

224464 (1S,2R,5S)-(+)-Menthol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?