• USA Home
  • 63685 - (−)-Menthyloxyacetic acid

EMAIL THIS PAGE TO A FRIEND
63685 Sigma-Aldrich

(−)-Menthyloxyacetic acid

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (−)-Menthyl carboxymethyl ether

  • CAS Number 40248-63-3

  • Empirical Formula (Hill Notation) C12H22O3

  • Molecular Weight 214.30

  •  Beilstein Registry Number 2444474

  •  EC Number 254-857-2

  •  MDL number MFCD00001483

  •  PubChem Substance ID 329759890

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   CILPHQCEVYJUDN-OUAUKWLOSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D −93±3°, c = 10% in ethanol
optical purity   enantiomeric ratio: ≥98.25:1.75 (GC)
refractive index   n20/D 1.4672(lit.)
bp   163-164 °C/10 mmHg(lit.)
mp   52-55 °C(lit.)
density   1.01 g/mL at 20 °C(lit.)

Description

Other Notes

Reagent for the resolution of amines, amino acids, alcohols and phenols

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?