• USA Home
  • 63844 - 2-Mercaptopyridine N-oxide sodium salt

EMAIL THIS PAGE TO A FRIEND
63844 Sigma-Aldrich

2-Mercaptopyridine N-oxide sodium salt

≥96.0% (NT)

Synonym: 1-Hydroxy-2-pyridinethione sodium salt, 2-Mercaptopyridine-1-oxide sodium salt, 2-Pyridinethiol-1-oxide sodium salt, Pyrithione sodium salt

  • CAS Number 3811-73-2

  • Empirical Formula (Hill Notation) C5H4NNaOS

  • Molecular Weight 149.15

  •  Beilstein Registry Number 4026050

  •  EC Number 223-296-5

  •  MDL number MFCD01941547

Purchase

Description

Other Notes

Esters of carboxylic acids with this N-oxide undergo decarboxylative free radical chain reactions

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UT9000000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?