• USA Home
  • 63852 - (Chloromethyl)polystyrene

EMAIL THIS PAGE TO A FRIEND
63852 Sigma-Aldrich

(Chloromethyl)polystyrene

extent of labeling: ~1.1 mmol/g Cl loading

Synonym: Merrifield resin

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Resins for Boc-/Bzl-SPPS More...
extent of labeling   ~1.1 mmol/g Cl loading
matrix   crosslinked with 1% DVB
particle size   200-400 mesh

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?