EMAIL THIS PAGE TO A FRIEND
63911 Sigma-Aldrich

Mesitylene

purum, ≥98.0% (GC)

Synonym: 1,3,5-Trimethylbenzene

Purchase

Properties

vapor density   4.1 (vs air)
vapor pressure   14 mmHg ( 55 °C)
  2.49 mmHg ( 25 °C)
InChI Key   AUHZEENZYGFFBQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   1022 °F
expl. lim.   0.88-6.1 %, 100 °F
refractive index   n20/D 1.499(lit.)
  n20/D 1.499
bp   163-166 °C(lit.)
mp   −45 °C(lit.)
density   0.864 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 2325 3 / PGIII
WGK Germany 
2
RTECS 
OX6825000
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M7200 Mesitylene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?