EMAIL THIS PAGE TO A FRIEND
64090 Sigma-Aldrich

Methacrylamide

purum, ≥98.0% (HPLC)

  • CAS Number 79-39-0

  • Linear Formula H2C=C(CH3)CONH2

  • Molecular Weight 85.10

  •  Beilstein Registry Number 605397

  •  EC Number 201-202-3

  •  MDL number MFCD00008018

  •  PubChem Substance ID 24882665

Purchase

Properties

Related Categories Amides, Building Blocks, C2 to C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   FQPSGWSUVKBHSU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
contains   stabilizer
mp   106-110 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UC6475000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?