EMAIL THIS PAGE TO A FRIEND
64151 Sigma-Aldrich

Lauryl methacrylate

purum, ≥95.0% (GC)

Synonym: Dodecyl methacrylate

  • CAS Number 142-90-5

  • Linear Formula CH2=C(CH3)COO(CH2)11CH3

  • Molecular Weight 254.41

  •  Beilstein Registry Number 1708160

  •  EC Number 205-570-6

  •  MDL number MFCD00008972

  •  PubChem Substance ID 329759931

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   GMSCBRSQMRDRCD-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
contains   hydroquinone monomethyl ether as stabilizer
refractive index   n20/D 1.445(lit.)
  n20/D 1.445
bp   142 °C/4 mmHg(lit.)
mp   −7 °C(lit.)
density   0.873 g/mL at 20 °C
  0.868 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
1
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

291811 Lauryl methacrylate, contains 500 ppm MEHQ as inhibitor, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?