• USA Home
  • 64161 - Glycidyl methacrylate

EMAIL THIS PAGE TO A FRIEND
64161 Sigma-Aldrich

Glycidyl methacrylate

purum, ≥97.0% (GC)

Synonym: 2,3-Epoxypropyl methacrylate, Methacrylic acid 2,3-epoxypropyl ester

  • CAS Number 106-91-2

  • Empirical Formula (Hill Notation) C7H10O3

  • Molecular Weight 142.15

  •  Beilstein Registry Number 2506

  •  EC Number 203-441-9

  •  MDL number MFCD00005137

  •  PubChem Substance ID 24883127

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   VOZRXNHHFUQHIL-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
contains   ~0.005% hydroquinone monomethyl ether as stabilizer
refractive index   n20/D 1.449(lit.)
  n20/D 1.450
bp   189 °C(lit.)
density   1.075 g/mL at 20 °C
  1.042 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Target organs 
Respiratory Tract
RIDADR 
UN 2922 6.1(8) / PGIII
WGK Germany 
1
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?