• USA Home
  • 64292 - Ethyl methanesulfonate

EMAIL THIS PAGE TO A FRIEND
64292 Sigma-Aldrich

Ethyl methanesulfonate

≥98.0% (T)

Synonym: Ethyl mesylate, Methanesulfonic acid ethyl ester

  • CAS Number 62-50-0

  • Linear Formula CH3SO3CH2CH3

  • Molecular Weight 124.16

  •  Beilstein Registry Number 773969

  •  EC Number 200-536-7

  •  MDL number MFCD00007559

  •  PubChem Substance ID 329759950

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonates/Sulfinates, Sulfur Compounds More...
InChI Key   PLUBXMRUUVWRLT-UHFFFAOYSA-N
assay   ≥98.0% (T)
refractive index   n20/D 1.418(lit.)
bp   85-86 °C/10 mmHg(lit.)
density   1.206 g/mL at 20 °C

Description

Other Notes

Mutagen. Mutagenesis of Chinese hamster ovary (CHO) cells

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
PB2100000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles

Articles

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?