EMAIL THIS PAGE TO A FRIEND
64330 Sigma-Aldrich

D-Methionine

puriss., ≥99.0% (NT)

Synonym: (R)-2-Amino-4-(methylmercapto)butyric acid, D-2-Amino-4-(methylthio)butanoic acid

  • CAS Number 348-67-4

  • Empirical Formula (Hill Notation) C5H11NO2S

  • Molecular Weight 149.21

  •  Beilstein Registry Number 1722293

  •  EC Number 206-483-6

  •  MDL number MFCD00002622

  •  PubChem Substance ID 329759958

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Methionine, Peptide Chemistry More...
InChI Key   FFEARJCKVFRZRR-SCSAIBSYSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D −23.7±0.5°, c = 5% in 5 M HCl

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
PD0455000
Protocols & Articles

Articles

HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M9375 D-Methionine, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?