• USA Home
  • 64701 - 3′-Methoxyacetophenone

EMAIL THIS PAGE TO A FRIEND
64701 Sigma-Aldrich

3′-Methoxyacetophenone

purum, ≥95.0% (GC)

Synonym: 3-Acetylanisole

  • CAS Number 586-37-8

  • Linear Formula CH3OC6H4COCH3

  • Molecular Weight 150.17

  •  Beilstein Registry Number 878552

  •  EC Number 209-573-3

  •  MDL number MFCD00008736

  •  PubChem Substance ID 329759993

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   BAYUSCHCCGXLAY-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.542(lit.)
  n20/D 1.542
bp   239-241 °C(lit.)
density   1.094 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M9408 3′-Methoxyacetophenone, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?