• USA Home
  • 64732 - 2-[2-(2-Methoxyethoxy)ethoxy]acetic acid

EMAIL THIS PAGE TO A FRIEND
64732 Sigma-Aldrich

2-[2-(2-Methoxyethoxy)ethoxy]acetic acid

technical, ≥90% (GC)

Synonym: O-[2-(2-Methoxyethoxy)ethyl]glycolic acid

  • CAS Number 16024-58-1

  • Linear Formula CH3(OCH2CH2)2OCH2CO2H

  • Molecular Weight 178.18

  •  Beilstein Registry Number 1769137

  •  EC Number 240-162-1

  •  MDL number MFCD00044098

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Functionalized Oligoethylene Glycol, PEGylation
InChI Key   YHBWXWLDOKIVCJ-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.446(lit.)
  n20/D 1.446
bp   140 °C/2 mmHg(lit.)
density   1.161 g/mL at 25 °C(lit.)

Description

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
1
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Articles

Peptide PEGylation

Circulatory half-life is a key success factor of new drugs. In this respect, modification of potential candidates ranging from non-peptidic molecules to peptides and proteins with polyethyleneglycol ...
Matthias Junkers
Aldrich ChemFiles 2007, 7.2, 15.
Keywords: Degradations, Drug discovery, Eliminations, Help, Reductions

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

407003 2-[2-(2-Methoxyethoxy)ethoxy]acetic acid, technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?