EMAIL THIS PAGE TO A FRIEND
64780 Sigma-Aldrich

m-Anisaldehyde

purum, ≥97.0% (GC)

Synonym: 3-Methoxybenzaldehyde

  • CAS Number 591-31-1

  • Linear Formula CH3OC6H4CHO

  • Molecular Weight 136.15

  •  Beilstein Registry Number 606013

  •  EC Number 209-712-8

  •  MDL number MFCD00003361

  •  PubChem Substance ID 24883551

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C8, Carbonyl Compounds, Chemical Synthesis,
InChI Key   WMPDAIZRQDCGFH-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.553(lit.)
  n20/D 1.553
bp   100-103 °C/10 mmHg
  143 °C/50 mmHg(lit.)
density   1.117 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BZ2605000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?