• USA Home
  • 64941 - Methyl (triphenylphosphoranylidene)acetate

EMAIL THIS PAGE TO A FRIEND
64941 Sigma-Aldrich

Methyl (triphenylphosphoranylidene)acetate

purum, ≥97.0% (CH)

Synonym: (Carbomethoxymethylene)triphenylphosphorane, (Methoxycarbonylmethylene)triphenylphosphorane

  • CAS Number 2605-67-6

  • Linear Formula (C6H5)3P=CHCO2CH3

  • Molecular Weight 334.35

  •  Beilstein Registry Number 618430

  •  EC Number 220-018-4

  •  MDL number MFCD00008455

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Olefination, Synthetic Reagents, Wittig Reagents More...
InChI Key   NTNUDYROPUKXNA-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (CH)
mp   168-172 °C(lit.)
  168-172 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

157929 Methyl (triphenylphosphoranylidene)acetate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?