650501 Sigma-Aldrich

Acetone

CHROMASOLV® Plus, for HPLC, ≥99.9%

DOWNLOAD MSDS (PDF)

Properties

Related Categories Acetone, Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV Plus,
grade   CHROMASOLV® Plus
  for HPLC
vapor density   2 (vs air)
vapor pressure   184 mmHg ( 20 °C)
assay   ≥99.9%
expl. lim.   13.2 %
impurities   ≤0.5% water
  ≤1.0 ppb Fluorescence (quinine) at 365 nm
evapn. residue   ≤0.0003%
halogenated residue   <10 ng/L (as heptachlor epoxide)
refractive index   n20/D 1.359(lit.)
bp   56 °C/760 mmHg(lit.)
mp   −94 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 330 nm Amax: 1.0
  λ: 340 nm Amax: 0.06
  λ: 350 nm Amax: 0.01
  λ: 375-400 nm Amax: 0.005

Description

Application

Suitable for HPLC, spectrophotometry, environmental testing

Acetone′s luminesence intensity is dependent upon the solution components 1. The absorption of UV light by acetone, results in its photolysis and the production of radials 1.

Packaging

1, 6×1, 4, 4×4 L in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Acetone
270725Acetone

CHROMASOLV®, for HPLC, ≥99.9%

Acetone
34850Acetone

CHROMASOLV®, for HPLC, ≥99.8%

Acetone
439126Acetone

CHROMASOLV®, for HPLC, ≥99.9%

Acetone
414689Acetone

capillary GC grade, ≥99.9%

Ethanol
E7023Ethanol

200 proof (absolute), for molecular biology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?