• USA Home
  • 65060 - 5-Methoxy-3-indoleacetic acid

EMAIL THIS PAGE TO A FRIEND
65060 Sigma-Aldrich

5-Methoxy-3-indoleacetic acid

purum, ≥99.0% (T)

Synonym: 2-(5-Methoxy-3-indolyl)acetic acid

  • CAS Number 3471-31-6

  • Empirical Formula (Hill Notation) C11H11NO3

  • Molecular Weight 205.21

  •  Beilstein Registry Number 187161

  •  EC Number 222-438-3

  •  MDL number MFCD00005638

  •  PubChem Substance ID 329760031

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   COCNDHOPIHDTHK-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   145-148 °C (dec.)(lit.)

Description

Application

• Reactant for preparation of aryloxybenzothiazoles as inhibitors of NO production
• Reactant for preparation of transcription inhibitors of novel Gli1 as antitumor agents
• Reactant for preparation of indole derivatives as NR2B/NMDA receptor antagonists
• Reactant for preparation of indolyl esters and amides related to indomethacin as selective COX-2 inhibitors
• Reactant for preparation of quinoline salicylic acid series of P-selectin antagonists
• Reactant for preparation of prostaglandin D2 receptor antagonists

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?