• USA Home
  • 65170 - 4-(4-Methoxyphenyl)butyric acid

EMAIL THIS PAGE TO A FRIEND
65170 Sigma-Aldrich

4-(4-Methoxyphenyl)butyric acid

purum, ≥97.0% (T)

  • CAS Number 4521-28-2

  • Linear Formula CH3OC6H4(CH2)3CO2H

  • Molecular Weight 194.23

  •  Beilstein Registry Number 2416568

  •  EC Number 224-849-3

  •  MDL number MFCD00004404

  •  PubChem Substance ID 24883856

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   LZHMNCJMXQKSBY-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
mp   56-59 °C(lit.)
  58-61 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

163384 4-(4-Methoxyphenyl)butyric acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?