• USA Home
  • 65208 - (S)-(+)-α-Methoxyphenylacetic acid

EMAIL THIS PAGE TO A FRIEND
65208 Sigma-Aldrich

(S)-(+)-α-Methoxyphenylacetic acid

for chiral derivatization, ≥99.0%

Synonym: O-Methyl-L-mandelic acid

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chiral, Chiral Derivatization Reagents, Chiral LC Derivatization Reagents,
grade   for chiral derivatization
InChI Key   DIWVBIXQCNRCFE-QMMMGPOBSA-N
assay   ≥99.0% (T)
  ≥99.0%
optical activity   [α]20/D +149±2°, c = 0.5% in ethanol
optical purity   enantiomeric ratio: ≥99.5:0.5 (GC)
mp   64-66 °C(lit.)
  64-67 °C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Reactant involved in the synthesis of biologically active molecules including:
• Acyclonucleoside phosphonates structural analogs of adefovir
• Labeled discodermolide for studying binding to tubulin
• 10-Isocyano-4-cadinene used for antifouling activity

Reactant involved in studies of immunostimulating chromanones gonytolides A-C

Reactant involved in:
• Hydroxylations and epoxidations
• Hydrogenations

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?