• USA Home
  • 65230 - 3-(2-Methoxyphenyl)propionic acid

EMAIL THIS PAGE TO A FRIEND
65230 Sigma-Aldrich

3-(2-Methoxyphenyl)propionic acid

purum, ≥98.0% (T)

Synonym: 2-Methoxyhydrocinnamic acid

  • CAS Number 6342-77-4

  • Linear Formula CH3OC6H4CH2CH2CO2H

  • Molecular Weight 180.20

  •  Beilstein Registry Number 2097012

  •  EC Number 228-738-0

  •  MDL number MFCD00002772

  •  PubChem Substance ID 329760044

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   XSZSNLOPIWWFHS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   85-87 °C(lit.)
  85-88 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?