EMAIL THIS PAGE TO A FRIEND
65357 Sigma-Aldrich

4-Methoxythiophenol

purum, ≥98.0% (GC)

Synonym: 4-Methoxybenzenethiol, 4-Mercaptoanisole

  • CAS Number 696-63-9

  • Linear Formula CH3OC6H4SH

  • Molecular Weight 140.20

  •  Beilstein Registry Number 1446910

  •  EC Number 211-799-2

  •  MDL number MFCD00004849

  •  PubChem Substance ID 24883964

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
InChI Key   NIFAOMSJMGEFTQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.5831(lit.)
  n20/D 1.584
bp   100-103 °C/13 mmHg(lit.)
density   1.14 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
RTECS 
DC1800000
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

109525 4-Methoxythiophenol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?