EMAIL THIS PAGE TO A FRIEND
65430 Sigma-Aldrich

N-Methylacetamide

purum, ≥98.0% (GC)

Synonym: Acetylmethylamine

  • CAS Number 79-16-3

  • Linear Formula CH3CONHCH3

  • Molecular Weight 73.09

  •  Beilstein Registry Number 1071255

  •  EC Number 201-182-6

  •  MDL number MFCD00008683

  •  PubChem Substance ID 329760004

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Protected Amines More...
InChI Key   OHLUUHNLEMFGTQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.433(lit.)
bp   204-206 °C(lit.)
mp   26-28 °C(lit.)
density   0.957 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AC5960000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M26305 N-Methylacetamide, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?